ContactUsLinkComponent
Filter Plate, Acropep Advance 96-well, 350 uL, 0.45 um Supor membrane, Height (Metriccm)1.8cm, Length (Metriccm): 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene, can be used in a number of high throughput applications including aqueous filtration, protein purification
Catalog #: MPCA97052-098
Supplier:  Cytiva
Filter Plate, Acropep Advance 96-well, 350 uL, 0.45 um Supor membrane, Height (Metriccm)1.8cm, Length (Metriccm): 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene, can be used in a number of high throughput applications including aqueous filtration, protein purification
Catalog #: MPCA97052-098
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.