ContactUsLinkComponent
Filter Plate, AcroPrep Advance, for DNA Purification 350uL, DNA binding media that ensures high yields of contaminant-free DNA for downstream applications, such as PCR and sequencing, Height (Metriccm)1.8cm, Length (Metriccm): 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89137-182
Supplier:  Cytiva
Filter Plate, AcroPrep Advance, for DNA Purification 350uL, DNA binding media that ensures high yields of contaminant-free DNA for downstream applications, such as PCR and sequencing, Height (Metriccm)1.8cm, Length (Metriccm): 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89137-182
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.