ContactUsLinkComponent
Filter Plate, AcroPrep Advance 96-Well, With Mustang Q/S Ion Exchange Membrane - 1 mL, Mustang Q membrane, can be used in high throughput ion exchange (IEX) applications, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89135-704
Supplier:  Cytiva
Filter Plate, AcroPrep Advance 96-Well, With Mustang Q/S Ion Exchange Membrane - 1 mL, Mustang Q membrane, can be used in high throughput ion exchange (IEX) applications, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89135-704
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.