ContactUsLinkComponent
Filter Plate, AcroPrep Advance, for Solvent Filtration 1 mL, 0.2 um PTFE membrane, can be used for high-throughput sample preparation of samples using solvents, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89135-692
Supplier:  Cytiva
Filter Plate, AcroPrep Advance, for Solvent Filtration 1 mL, 0.2 um PTFE membrane, can be used for high-throughput sample preparation of samples using solvents, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89135-692
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.