ContactUsLinkComponent
Filter Plate, AcroPrep Advance 96-Well, 1 mL, 1.0 um Glass Fiber, can be used in a number of aqueous high throughput filtration applications including general sample preparation, Height (Metriccm)1.8cm, Length (Metriccm): 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89135-688
Supplier:  Cytiva
Filter Plate, AcroPrep Advance 96-Well, 1 mL, 1.0 um Glass Fiber, can be used in a number of aqueous high throughput filtration applications including general sample preparation, Height (Metriccm)1.8cm, Length (Metriccm): 12.8cm, Width (Metric): 8.6cm, Plate Housing Material: Polypropylene
Catalog #: MPCA89135-688
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.