ContactUsLinkComponent
Filter plate, AcroPrep Advance 96-well, with0.2um next generation universal membrane, Filter Media: WWPTFE (water-wettable polytetrafluoroethylene) membrane
Catalog #: MPCA76308-650
Supplier:  Cytiva
Filter plate, AcroPrep Advance 96-well, with0.2um next generation universal membrane, Filter Media: WWPTFE (water-wettable polytetrafluoroethylene) membrane
Catalog #: MPCA76308-650
Supplier:  Cytiva
Customers Who Bought This Also Bought