Filter plate, AcroPrep Advance 96-well, with 0.2um next generation universal membrane, Filter Media: WWPTFE (water-wettable polytetrafluoroethylene) membrane
Catalog #: MPCA76308-646
Supplier: Cytiva
Filter plate, AcroPrep Advance 96-well, with 0.2um next generation universal membrane, Filter Media: WWPTFE (water-wettable polytetrafluoroethylene) membrane
Catalog #: MPCA76308-646
Supplier: Cytiva
Customers Who Bought This Also Bought
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
1 747,88 $
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
362,33 $
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
209,41 $ to 1 276,12 $
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
534,61 $ to 1 523,31 $