ContactUsLinkComponent
Filter Plate, AcroPrep Advance 96-well Long Tip, for Nucleic Acid Binding - Long Tip Filter Plate for Nucleic Acid Binding, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA10158-728
Supplier:  Cytiva
Filter Plate, AcroPrep Advance 96-well Long Tip, for Nucleic Acid Binding - Long Tip Filter Plate for Nucleic Acid Binding, Height (Metric cm)1.8 cm, Length (Metric cm): 12.8 cm, Width (Metric): 8.6 cm, Plate Housing Material: Polypropylene
Catalog #: MPCA10158-728
Supplier:  Cytiva
Customers Who Bought This Also Bought