Gloves, Edge 48-706VP, 13G Grey HPPE (Nylon, Polyester, Spandex, Glass Fiber) Knit Liner, Black Foam Nitrile Palm Coat, Elasticized Knitwrist Cuff, Vend Pack, Size: 6
Catalog #: CA76519-388
Supplier: Ansell Healthcare
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.