ContactUsLinkComponent
Syringe Filter, Filter Media: wwPTFE, For HPLC/UHPLC Sample Preparation, constructed with solvent compatible polypropylene material, designed to protect analytical instrumentation and improve the reliability of results, Pore Size: 0.2Um, Filter Size: 13Mm
Catalog #: MPCA76308-728
Supplier:  Cytiva
Syringe Filter, Filter Media: wwPTFE, For HPLC/UHPLC Sample Preparation, constructed with solvent compatible polypropylene material, designed to protect analytical instrumentation and improve the reliability of results, Pore Size: 0.2Um, Filter Size: 13Mm
Catalog #: MPCA76308-728
Supplier:  Cytiva
Customers Who Bought This Also Bought