ContactUsLinkComponent
Syringe Filter, Minipike outlet, Filter Media: wwPTFE, For HPLC/UHPLC Sample Preparation, constructed with solvent compatible polypropylene material, designed to protect analytical instrumentation and improve the reliability of results, Pore size: 0.45 Um, Filter size: 13Mm
Catalog #: MPCA76308-700
Supplier:  Cytiva
Syringe Filter, Minipike outlet, Filter Media: wwPTFE, For HPLC/UHPLC Sample Preparation, constructed with solvent compatible polypropylene material, designed to protect analytical instrumentation and improve the reliability of results, Pore size: 0.45 Um, Filter size: 13Mm
Catalog #: MPCA76308-700
Supplier:  Cytiva
Customers Who Bought This Also Bought