ContactUsLinkComponent
Syringe Filter, Psf, GxF/wwPTFE membrane, features GxF multi-layer glass fiber prefilter, providing two to four times the throughput of standard prefilter devices, multi-layer prefilter captures particulate rated from > 40 to 1 um, For HPLC/UHPLC Sample Preparation, Size: 25Mm, Pore Size: 0.45Um
Catalog #: MPCA76308-680
Supplier:  Cytiva
Syringe Filter, Psf, GxF/wwPTFE membrane, features GxF multi-layer glass fiber prefilter, providing two to four times the throughput of standard prefilter devices, multi-layer prefilter captures particulate rated from > 40 to 1 um, For HPLC/UHPLC Sample Preparation, Size: 25Mm, Pore Size: 0.45Um
Catalog #: MPCA76308-680
Supplier:  Cytiva
Customers Who Bought This Also Bought