Order Entry
ContactUsLinkComponent
Inlet septa, 11 mm, Non-stick Advanced Green, 50/pk
Advanced Green Septa for GC, Agilent Technologies
Catalog #: CAAG8010-0207
Inlet septa, 11 mm, Non-stick Advanced Green, 50/pk
Advanced Green Septa for GC, Agilent Technologies
Catalog #: CAAG8010-0207

Specifications

  • Ø:
    11 mm
  • Maximum Temperature:
    350 °C
  • Description:
    Inlet septa, non-stick
  • Material:
    Silicone
  • Cat. no.:
    CAAG8010-0207
  • Type:
    Advanced Green

Specifications

About this item

Advanced green nonstick 11 mm septa eliminates sticking leading to scraping bits of spetum out of the GC inlet.

  • Our green septa provide more injections per septum
  • Plasma coating eliminates sticking in the injection port
  • Maximum injection port temperature: 350 °C
  • Features blister packaging for cleanliness and convenience

Agilent now coats each premium 11 mm inlet septa with plasma to eliminate sticking in the GC inlet. With our premium non-stick septa, you'll never have to scrape bits of septum out of your inlet. Our premium septa also have a recess on the injection side that guides the syringe needle to the same point with every injection.

Customers Who Bought This Also Bought