ContactUsLinkComponent
Pipette Filler, Bulb, Universal, Synthetic Rubber, Red/White, with 3 glass ball valves that are controlled by finger pressure, Comes complete with adapter for pipettes with capacities greater than 20mL, no metal parts to corrode and the filler can be used with most common solutions
Catalog #: 76519-482
Supplier:  Globe Scientific
Pipette Filler, Bulb, Universal, Synthetic Rubber, Red/White, with 3 glass ball valves that are controlled by finger pressure, Comes complete with adapter for pipettes with capacities greater than 20mL, no metal parts to corrode and the filler can be used with most common solutions
Catalog #: 76519-482
Supplier:  Globe Scientific
Customers Who Bought This Also Bought