VWR® Nitrile Examination Gloves, Blue
Catalog #: 76518-342
Supplier: VWR International
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue
- Glove size:XL
- Length:22.9 cm (9")
- Powdered/Powder-free:Powder-free
- Latex/Latex-free:Latex-free
- Material:Nitrile
- Sterility:Non sterile
- Cuff style:Beaded
- Finish:Textured fingers
Specifications
About this item
Ambidextrous gloves are soft and comfortable for everyday use. They protect hands from a broad range of chemicals. Beaded cuff allows easy donning.
- Finger textured
- Ambidextrous
- Nonsterile
- Soft and comfortable
- Protects hands from broad range of chemicals and hazards
- Powder-free
Ideal for use in industrial and laboratory applications.
Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Finger textured gloves feature rolled, beaded cuffs.
Certifications: Meet or exceed ASTM D6319 on water leak and dimension. Manufactured under GMP, ISO9001, ISO13485 and MDSAP quality system.
Packaging: 100 gloves per box, 10 boxes per case.
Customers Who Bought This Also Bought