ContactUsLinkComponent
SPS 4 probe 0.8 mm id with green markers. Inert, FEP sleeved for SPS 4 autosampler
CrossLab Autosampler Probes, Agilent Technologies
Catalog #: CAG8410-80102
SPS 4 probe 0.8 mm id with green markers. Inert, FEP sleeved for SPS 4 autosampler
CrossLab Autosampler Probes, Agilent Technologies
Catalog #: CAG8410-80102
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    SPS 4 probe 0.8 mm Int. Ø with green markers. Inert, FEP sleeved for SPS 4 autosampler
  • Cat. no.:
    CAG8410-80102
Specifications
About this item
Sampling probes for autosamplers used with Agilent AAS, ICP-OES, MP-AES, and ICP-MS instruments.
  • Compatible with Agilent atomic spectroscopy instruments: AAS, MP-AES, ICP-OES and ICP-MS
  • High-speed probe arm assembly and optimized movement for fast sample-to-sample speed
  • Multiple probe size options allows you to select the right probe for your application
  • PTFE or FEP probes and capillaries reduce contamination in the instrument
  • Easy replacement by the user
  • Optional SPS 4 probe available to support sampling from 96-well microplates (with Agilent ICP-MS only)
Many types of autosamplers have been used with Agilent AA, ICP-OES, MP-AES and ICP-MS instruments, depending on the specific model. This includes the Agilent SPS 4 used with current instruments, and the SPS 3 and ASX 500 series used with earlier model instruments. Replacement autosampler probes for the Agilent SPS 4, SPS 3 and the Agilent ASX 500 series autosamplers are available.
Dispensing capillary assemblies are made from PTFE or FEP tubing, making them ideal for collecting a variety of corrosive matrices and organic solutions;Works well with solvent and acid-resistant sampling components.
Customers Who Bought This Also Bought