CrossLab Skimmer Cones for ICP-MS, Agilent Technologies
Catalog #: CAG8400-67201
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:7900, 8900 with x-lens ICP-MS skimmer cone, Platinum with Copper base. Optional cone used with x-lens for aggressive acid digests, including HF
- Cat. no.:CAG8400-67201
Specifications
About this item
Agilent skimmer cones ensure the performance that single & triple quadrupole ICP-MS systems demand.
- Nickel skimmer cones are suitable for most common applications and provide the most economical operation
- Platinum skimmer cones are required for analysis of aggressive acids (especially HF) and when O2/Ar option gas is used for analysis of organic solvents
- Platinum skimmer with copper base is recommended for the lowest LODs and for higher matrix samples
- Platinum skimmer with nickel base is recommended for organics analysis
Agilent, nickel or platinum tipped skimmer cones use high-purity materials to minimize background and ensure stability within the plasma. Cones are typically the biggest supplies cost in routine ICP-MS operation. Protect your investment and ensure ease of use by choosing genuine Agilent cones.High-quality interface cones are key to the sensitivity and stability of an ICP-MS. Our skimmer cones are manufactured to stringent specifications and rigorously tested to ensure excellent batch to batch reproducibility. The orifice dimensions and tip geometry are optimized and tightly controlled.
Customers Who Bought This Also Bought