Closures for High Recovery Vials, Agilent Technologies
Catalog #: CAG6500-88027
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Closures for High Recovery Vials, Agilent Technologies
Catalog #: CAG6500-88027
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Snap caps and seals for 10 ml wash vials
- Cat. no.:CAG6500-88027
Specifications
About this item
Customers Who Bought This Also Bought
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$99.85 to $385.36
Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
$145.16 to $1,345.99
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$260.48 to $788.04
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
$46.00 to $367.00
Hydrofluoric acid 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$311.43