ContactUsLinkComponent
Kit, Vi-Cell Blu Reagent, Ready-to-load, includes trypan blue, waste bottle, buffer solution, disinfectant and cleaning agent. Each pack can be used for 310 activities before changing and includes an RFID tag showing how many activities are remaining, lot number, expiration date, and on-board life
Vi-CELL BLU Reagent Kits, Beckman Coulter
Catalog #: CA76409-827
Supplier:  Beckman Coulter
CAS Number:  
Kit, Vi-Cell Blu Reagent, Ready-to-load, includes trypan blue, waste bottle, buffer solution, disinfectant and cleaning agent. Each pack can be used for 310 activities before changing and includes an RFID tag showing how many activities are remaining, lot number, expiration date, and on-board life
Vi-CELL BLU Reagent Kits, Beckman Coulter
Catalog #: CA76409-827
Supplier:  Beckman Coulter
CAS Number:  
Specifications
  • Description:
    Vi-CELL BLU reagent kit
Specifications
About this item
We have improved the Vi-CELL reagent pack to make it easier to use. The Vi-CELL reagents you know and trust are now packaged in a convenient ready-to-load kit. Simply open the instrument reagent panel and slide in the reagent pack. Each pack can be used for 310 activities before changing. Packs are easy to replace with no mess. Say goodbye to wrangling with tubes and splattering dye when you change out reagents.
Each pack also includes an RFID tag that tells you how many activities are remaining on the pack. No more guessing on remaining reagent amounts or running out in the middle of a critical series of tests.
The embedded RFID tag gives important information about each pack to your Vi-CELL BLU instrument: Lot number, expiration date, on-board life (90 days), activities remaining and part number.
Each reagent pack contains: Trypan blue bottle, waste bottle, buffer solution bottle, disinfectant bottle and cleaning agent bottle.
Ordering information: C06019 is for a single Vi-CELL BLU Reagent pack. C39291 is for a quad pack (Quantity 4) of Vi-CELL BLU reagent packs.
Customers Who Bought This Also Bought