Specifications
- Volume:500 µl
- Description:Manual syringe, PTFE tipped plunger
- Cat. no.:CA5190-1523
- Needle Length:50 mm
- Needle Type:Fixed
- Needle Gauge:25
- Tip Type:Bevel
Specifications
About this item
Agilent syringes for manual injection can be used with any type or brand of manual injection valve.
- Blunt-tip point style prevents damage to valve's internal parts
- Can be used with any type or brand of manual injection valve
Agilent syringes for manual injection have a blunt-tip needle to prevent damaging the valve's internal parts. These syringes can be used with any type or brand of manual injection valve. For more help choosing the right syringe for your application, check out our syringe selection tool.