Cleanroom Upgrade, For 7400-25 Aggressive, series incubator to handle exterior wipedown with aggressive cleaning chemicals, includes 316 stainless steel on all exterior surfaces as well as hinges and hardware, coated magnetic door trim, and capacitive touchscreen
Catalog #: 76518-144
Supplier: Caron Products
Customers Who Bought This Also Bought