Order Entry
ContactUsLinkComponent
Suction Flask, Color: Translucent, Includes 9mm Plastic safety hose nipple, For Use With (Application): For collecting the filtrate, Material: Borosilicate Glass, Stopper/tube, Sterilizable, Volume (Metric): 1L
Filtering Flasks, with Plastic Hose Connection, Sartorius
Catalog #: CA76517-778
Supplier:  Sartorius
Filtering Flasks, with Plastic Hose Connection, Sartorius
Catalog #: CA76517-778
Supplier:  Sartorius

Specifications

  • Capacity:
    1000 ml
  • Cat. no.:
    CA76517-778
  • Connection Size:
    Suction flask, neck Int. Ø.: 60 mm

Specifications

About this item

These vacuum-resistant filtering flasks made of DURAN® borosilicate glass, can be used in filtrate collection.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Standard according to DIN 12476.

Customers Who Bought This Also Bought