AMI Polymer Imaflow™ Platinum Cured Silicone Tubing, Foxx Life Sciences
Catalog #: 76516-840
Supplier: Foxx Life Sciences
Specifications
- Length:50 ft.
- Tubing I.D.:6.35 mm (¹/₄")
- Tubing O.D.:9.5 mm (³/₈")
- Color:Transparent
- Description:Platinum cured silicone tubing
- Cat. no.:76516-840
Specifications
About this item
Imaflow™ is Platinum Grade Silicone Tubing designed for general purpose applications in within and outside of the Food and Pharmaceutical industries. Imaflow™ tubing is made from Pharmaceutical grade platinum cured silicone resin with a smooth bore finish compared to other peroxide grade tubing to reduce the risk of particle entrapment during fluid transfer.
- Meets pharma grade requirement of ROW and Domestic markets
- Greater Transparency and higher shelf life compare to other peroxide tubing
- Free of heavy metals and Hazardous substances
- Service Temperature Range: −60 to +230 °C
- Acute Systemic Toxicity Test (USP Class VI)
Imaflow™ meets all technical requirements related to the non-regulatory market and is excellent with peristaltic pump operation.
Certifications: Meets pharma grade requirement of FDA 21 CFR 177.2600; Manufactured and packaged in Class 10000 Clean Room; Intracutaneous Toxicity Test (USP Class VI).
Customers Who Bought This Also Bought