Sampler Cones and Care Packages for ICP-MS
Catalog #: CAG3280-67040
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:ICP-MS sampler cone, Nickel-tip (12 mm Ø) with Copper base, for x-lens;s-lens. Suitable for 77/78/7900 and 88/8900. Standard cone used with x-lens for most routine applications.
- Cat. no.:CAG3280-67040
Specifications
About this item
Agilent sampler cones and cone care packages ensure ICP-MS system stability and long lifetime.
- Nickel tipped sampler cones are suitable for most common applications and provide the most economical operation
- Nickel tipped, nickel plated sampler cones are recommended for samples containing >0.5% HCl, or for routine operation with (U)HMI
- Platinum tipped sampler cones are required for analysis of aggressive acids (especially HF) and when O₂/Ar option gas is used for analysis of organic solvents
- Platinum tipped, nickel plated sampler cones are recommended for analysis of aggressive acids (especially HCl, HF, and HClO₄) and for higher-matrix samples
- Platinum tipped sampler cones with large 18 mm insert are recommended for for high-viscosity and high-boiling-point acids (e.g. H2SO₄, H3PO₄), or for the lowest LODs with semiconductor applications
- Cone care kits are also available for all Agilent ICP-MS systems, which bundle all the supplies you need to replace and maintain your interface cones in one convenient package
Agilent nickel or platinum tipped sampler cones use high-purity materials to minimize background and ensure stability within the plasma. Cones are typically the biggest supplies cost in routine ICP-MS operation. Protect your investment and ensure ease of use by choosing genuine Agilent cones.
High-quality interface cones are key to the sensitivity and stability of an ICP-MS. Our sampler cones are manufactured to stringent specifications and rigorously tested to ensure excellent batch to batch reproducibility. The orifice dimensions and tip geometry are optimized and tightly controlled.
Customers Who Bought This Also Bought