CrossLab Nebulizers for ICP-MS, Agilent Technologies
Catalog #: CAG3266-80004
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Nebulizer, MicroMist, U-series with UniFit sample connector, standard with Agilent 7800/7850/7900 Series ICP-MS, 40 µm
- Cat. no.:CAG3266-80004
Specifications
About this item
Agilent has MicroMist and inert PFA concentric nebulizers to suit your ICP-MS application needs.
- Concentric nebulizers are routinely recommended for use with ICP-MS for all but the highest matrix samples which may cause the inner capillary to become clogged by particles or high concentrations of salt
- Two types of concentric nebulizer are available: Quartz (recommended for trace analysis of clean samples) and PFA (recommended for semi-conductor process chemicals - providing inertness to HF and self-aspiration)
- High TDS/matrix samples are tolerated by parallel path nebulizers such as the MiraMist available in PEEK or PTFE for inertness to high acid concentration and solvents
- A full range of connection accessories including U-series and Ezy Fit sample connectors, tubing, O-rings and other fittings are available for all models, plus nebulizer cleaning tools, manufactured to Agilent's high quality standards
- Ensure the longevity of your nebulizer through regular cleaning with a 5% HNO₃ (v/v) bath (do not use an ultrasonic bath)
- Replace the nebulizer when the capillary is broken or the nebulizer is damaged
Agilent has a range of nebulizers and supplies to best suit your ICP-MS application needs. From standard glass or inert PFA concentric nebulizers, robust nebulizers for high TDS samples, to low-flow and self-aspirating nebulizers for your ICP-MS.
MicroMist glass concentric nebulizers and a full range of connection accessories including U-series and Ezy Fit sample connectors, tubing, nebulizer cleaning tools, O-rings and other fittings are available for all models.
Customers Who Bought This Also Bought