Specifications
- Ø:13 mm
- Pore size:0.22 µm
- Sterility:Sterile
- Cat. no.:76516-622
- Packaging:Individually wrapped
- Membrane:SFCA
Specifications
About this item
Sterile Diamond® PureFlow™ syringe filters satisfy requirements of life science applications for sterilization of solutions or clarification of biological fluids. Sterile filters include a polypropylene (PP) microfiber pre-filter layer to improve filtration of particulate-laden fluids that are difficult to filter with single layer syringe filters.
- SFCA (Surfactant Free Cellulose Acetate) membrane
- Low protein binding, low extractables and leachables, and high flow rate
- Ideal for cell culture media & serum, immunological samples, and biological fluids
- pH range: 4 to 8
- Female Luer lock inlet/male Luer slip outlet
Performance: High flow rates and low absorption membranes for minimal sample loss. Filters have a color coded outer ring which allows easy identification of membrane and filter media type. Each unit is clearly marked with an identifying code to denote pore size and membrane material. The bonded outer ring on housing provides strength under higher working pressures for higher performance needs. The integral polypropylene pre-filter for high flow rates and fast filtration ensures efficiency.