Stirrer, Rt2 Basic Hotplate, 140 mm dia, Top Plate Material: Aluminum, Benefit from exceptionally strong magnet coupling power and a smooth-start stirring mechanism, dimensions (L x W x H): 290 x 161 x 100mm, Two threaded holes for support rods hold various kinds of devices, Voltage: 120V
Catalog #: MP10054-156
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought