Order Entry
ContactUsLinkComponent
Ferrule, 0.4 mm id, graphite, 0.05 to 0.25 mm column
CrossLab Graphite Ferrules for GC, Agilent Technologies
Catalog #: CA500-2114
Ferrule, 0.4 mm id, graphite, 0.05 to 0.25 mm column
CrossLab Graphite Ferrules for GC, Agilent Technologies
Catalog #: CA500-2114

Specifications

  • I.D.:
    0.4 mm
  • Description:
    Ferrule, 0.4 mm Int.Ø, 0.05 to 0.25 mm column
  • Material:
    Graphite
  • Cat. no.:
    CA500-2114

Specifications

About this item

Graphite ferrules can withstand temperatures as high as 450 °C. Their inherent softness makes them ideal for metal columns and allows for additional installation tolerances.

  • General purpose for capillary columns
  • Suitable for FID, NPD, and ECD
  • Recommended for high temperature and cool on-column applications
  • Material is free of sulfur and other contaminants
  • Can be reused or removed easily
  • Proper installation requires a finger-tight turn on the nut, then an additional 1/4 turn with a wrench
  • Not for use with MS or oxygen sensitive detectors

Graphite ferrules can withstand temperatures as high as 450 °C making them a top choice for high temperature applications. Their inherent softness and thermal expansion properties help to prevent column damage due to overtightening while maintaining a leak-free seal throughout temperature cycling. Agilent offers a comprehensive selection of 100% graphite ferrules made for use with a variety of GC columns. These ferrules are recommended for making leak-free connections between the GC column and injector port in high temperature applications.

Customers Who Bought This Also Bought