Order Entry
ContactUsLinkComponent
Vial rack, plastic, for 12 mm crimp top micro vials
Sample Containment Accessories, Agilent Technologies
Catalog #: CA76515-132
Vial rack, plastic, for 12 mm crimp top micro vials
Sample Containment Accessories, Agilent Technologies
Catalog #: CA76515-132

Specifications

  • Description:
    Vial rack, plastic, for 12 mm crimp top micro vials
  • Cat. no.:
    CA76515-132

Specifications

About this item

Securely hold 2 ml vials for table top sample prep or store up to 50, 2 ml vials.

  • Agilent 12 mm vial racks are ideal for holding 2 ml glass vials ( 2 ml screw vials, 2 ml crimp vials or 2 ml snap vials) for table top sample prep
  • Vial storage boxes are able to hold 50 × 2 ml vials instead which offer a cover when compared to an open vial rack
Customers Who Bought This Also Bought