ContactUsLinkComponent
ALS syringe, 10 uL, fixed needle, 23/42/cone
CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC, Agilent Technologies
Catalog #: CA76515-128
ALS syringe, 10 uL, fixed needle, 23/42/cone
CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC, Agilent Technologies
Catalog #: CA76515-128
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ALS syringe, 10 µl, fixed needle, 23/42/cone
  • Cat. no.:
    CA76515-128
Specifications
About this item
Agilent programmed temperature vaporizer inlets combine the benefits of split, splitless, and on-column inlets, resulting in less thermal degradation of labile compounds.
  • Perform large volume injections for low minimum detection limits
  • Get less thermal degradation of labile compounds
Agilent programmed temperature vaporizer inlets combine the benefits of split, splitless, and on-column inlets. The sample is usually injected into a cool liner, so syringe needle discrimination does not occur. Then the inlet temperature is increased to vaporize the sample. You then program vent times and temperature to achieve the equivalent of split or splitless transfer of sample vapors to the column. PTV injection is considered the most universal sample introduction system because of its flexibility.
Customers Who Bought This Also Bought