ContactUsLinkComponent
Heater For Fractron 100, 1 zone of 18 kW of heating capacity. Pump Capacity: 1 HP, 18 GPM 30 PSI. 230 V, 1/8 DIN Controller - 4-20MA/0--10V, Zone 1. CE Certification available. Requires two 55-gal drums of Paratherm NF oil (Not included)
Catalog #: 76514-410
Supplier:  United Science
Heater For Fractron 100, 1 zone of 18 kW of heating capacity. Pump Capacity: 1 HP, 18 GPM 30 PSI. 230 V, 1/8 DIN Controller - 4-20MA/0--10V, Zone 1. CE Certification available. Requires two 55-gal drums of Paratherm NF oil (Not included)
Catalog #: 76514-410
Supplier:  United Science
Customers Who Bought This Also Bought