ContactUsLinkComponent
Kit Ethanol Co-Solvent Pump, for delivery of liquid solvent to the Extrakt 110, 140 or 180. Includes inert gas feed pump for drum feed tank delivery. Must be ordered with a new system purchase. System will be upgraded to UL698A for C1D2 compliancy.
Catalog #: 76514-408
Supplier:  United Science
Kit Ethanol Co-Solvent Pump, for delivery of liquid solvent to the Extrakt 110, 140 or 180. Includes inert gas feed pump for drum feed tank delivery. Must be ordered with a new system purchase. System will be upgraded to UL698A for C1D2 compliancy.
Catalog #: 76514-408
Supplier:  United Science
Customers Who Bought This Also Bought