ContactUsLinkComponent
Tobacco Etch Virus Protease, Source: E. Coli, Purity: > 95% by SDS-PAGE analyses, Molecular Weight: 28.4kDa, observed by reducing SDS-PAGE.Storage: Recombinant Tobacco Etch Virus Protease remains stable up to 1year at -20C from date of receipt, Synonyms: rTEV, TEV, P1 protease, Size: 10000 IU
Catalog #: 76512-418
Supplier:  Genscript
Tobacco Etch Virus Protease, Source: E. Coli, Purity: > 95% by SDS-PAGE analyses, Molecular Weight: 28.4kDa, observed by reducing SDS-PAGE.Storage: Recombinant Tobacco Etch Virus Protease remains stable up to 1year at -20C from date of receipt, Synonyms: rTEV, TEV, P1 protease, Size: 10000 IU
Catalog #: 76512-418
Supplier:  Genscript
Customers Who Bought This Also Bought