ContactUsLinkComponent
Biotin widely conjugated to proteins antibodies for biochemical assays. Avidin (streptavidin)-biotin system commonly used for many immunoassays such as ELISA, Flow Cytometry, Immunofluorescence, In Situ Hybridization, Immunohistochemistry. Biotin mAb better alternative to avi, Size: 10mg
Catalog #: 76512-248
Supplier:  Genscript
Biotin widely conjugated to proteins antibodies for biochemical assays. Avidin (streptavidin)-biotin system commonly used for many immunoassays such as ELISA, Flow Cytometry, Immunofluorescence, In Situ Hybridization, Immunohistochemistry. Biotin mAb better alternative to avi, Size: 10mg
Catalog #: 76512-248
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.