ContactUsLinkComponent
MSH6, also known as MutS protein homolog 6, is a member of the Mutator S (MutS) family of proteins. It functions to repair mutations during DNA replication. MSH6 is a useful in diagnostics of gastrointestinal tract cancer, Size: 10mg
Catalog #: 76512-236
Supplier:  Genscript
MSH6, also known as MutS protein homolog 6, is a member of the Mutator S (MutS) family of proteins. It functions to repair mutations during DNA replication. MSH6 is a useful in diagnostics of gastrointestinal tract cancer, Size: 10mg
Catalog #: 76512-236
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.