ContactUsLinkComponent
Thyroid stimulating hormone (TSH) is a pituitary hormone that stimulates thyroid gland to secrete thyroxine (T4) and triiodothyronine (T3). It is a glycoprotein, consisting of alpha and beta subunit. TSH serves as a sensitive and specific biomarker for evaluating thyroid function. It is, Size: 100mg
Catalog #: 76512-230
Supplier:  Genscript
Thyroid stimulating hormone (TSH) is a pituitary hormone that stimulates thyroid gland to secrete thyroxine (T4) and triiodothyronine (T3). It is a glycoprotein, consisting of alpha and beta subunit. TSH serves as a sensitive and specific biomarker for evaluating thyroid function. It is, Size: 100mg
Catalog #: 76512-230
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.