ContactUsLinkComponent
CA72-4 (Cancer antigen 72-4), also known as Tumor associated glycoprotein (TAG) 72, mucin-like protein of high molecular weight (200- 400 kD). CA 72-4 has high specificity for diagnos of benign diseases. It also a helpful marker in diagnos of gastric cancer, ovarian carcinoma col, Size: 100mg
Catalog #: 76512-212
Supplier:  Genscript
CA72-4 (Cancer antigen 72-4), also known as Tumor associated glycoprotein (TAG) 72, mucin-like protein of high molecular weight (200- 400 kD). CA 72-4 has high specificity for diagnos of benign diseases. It also a helpful marker in diagnos of gastric cancer, ovarian carcinoma col, Size: 100mg
Catalog #: 76512-212
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.