ContactUsLinkComponent
Glycated hemoglobin (HbA1c) is formed by hemoglobin's exposure to plasma glucose in a non-enzymatic process. As average amount of plasma glucose increases, fraction of HbA1c scales up. HbA1c reflects average plasma glucose over previous eight to twelve weeks it can be used as a marke, Size: 100mg
Catalog #: 76512-184
Supplier:  Genscript
Glycated hemoglobin (HbA1c) is formed by hemoglobin's exposure to plasma glucose in a non-enzymatic process. As average amount of plasma glucose increases, fraction of HbA1c scales up. HbA1c reflects average plasma glucose over previous eight to twelve weeks it can be used as a marke, Size: 100mg
Catalog #: 76512-184
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.