ContactUsLinkComponent
Glycated hemoglobin (HbA1c) is formed by hemoglobin's exposure to plasma glucose in a non-enzymatic process. As average amount of plasma glucose increases, fraction of HbA1c scales up. HbA1c reflects average plasma glucose over previous eight to twelve weeks and it can be used as a marke, Size: 10mg
Catalog #: 76512-170
Supplier:  Genscript
Glycated hemoglobin (HbA1c) is formed by hemoglobin's exposure to plasma glucose in a non-enzymatic process. As average amount of plasma glucose increases, fraction of HbA1c scales up. HbA1c reflects average plasma glucose over previous eight to twelve weeks and it can be used as a marke, Size: 10mg
Catalog #: 76512-170
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.