ContactUsLinkComponent
Human chorionic gonadotropin (HCG) is a polypeptide hormone produced by the human placenta. HCG is a heterodimer, composed of an alpha and a beta subunit. The level of HCG is elevated in the serum of pregnant women. It is a useful marker for diagnosis of pregnancy, Size: 10mg
Catalog #: 76512-134
Supplier:  Genscript
Human chorionic gonadotropin (HCG) is a polypeptide hormone produced by the human placenta. HCG is a heterodimer, composed of an alpha and a beta subunit. The level of HCG is elevated in the serum of pregnant women. It is a useful marker for diagnosis of pregnancy, Size: 10mg
Catalog #: 76512-134
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.