ContactUsLinkComponent
Protein Induced by Vitamin K Absence or Antagonist-II (PIVKA-II), also known as Des-Y-carboxy-prothrombin (DCP), abnormal form of prothrombin. Normally, prothrombin's 10 glutamic acid residues (Glu) in Y-carboxyglutamic acid (Gla) domain at positions 6, 7, 14, 16, 19, 20,25, 26, 29 and, Size: 10mg
Catalog #: 76512-106
Supplier:  Genscript
Protein Induced by Vitamin K Absence or Antagonist-II (PIVKA-II), also known as Des-Y-carboxy-prothrombin (DCP), abnormal form of prothrombin. Normally, prothrombin's 10 glutamic acid residues (Glu) in Y-carboxyglutamic acid (Gla) domain at positions 6, 7, 14, 16, 19, 20,25, 26, 29 and, Size: 10mg
Catalog #: 76512-106
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.