ContactUsLinkComponent
Carcinoembroyonic antigen (CEA) is a glycoprotein, produced during embryonic and fetal period. High concentration of CEA in blood are observed in many patients with cancer of lung, liver, pancreas, breast, colon and prostate. Elevated CEA levels are related to stage of diseas, Size: 10mg
Catalog #: 76512-040
Supplier:  Genscript
Carcinoembroyonic antigen (CEA) is a glycoprotein, produced during embryonic and fetal period. High concentration of CEA in blood are observed in many patients with cancer of lung, liver, pancreas, breast, colon and prostate. Elevated CEA levels are related to stage of diseas, Size: 10mg
Catalog #: 76512-040
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.