Suppression of Tumorigenicity 2 (ST2), a member of Interleukin-1 receptor family, has two isoforms, one is soluble form (sST2) and other one is transmembrane form (ST2L). SST2, lacking transmembrane and intracellular domains, can prevent interaction between IL-33 and ST2L. It plays a, Size: 10mg
Catalog #: 76512-022
Supplier: Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.