ContactUsLinkComponent
CA125, known as cancer antigen 125, carcinoma antigen 125, and carbohydrate antigen 125, is a member of the mucin family glycoproteins. It is involved in boosting tumorigenesis and tumor proliferation and is considered as a useful biomarker for the diagnosis of ovarian cancer, Size: 10mg
Catalog #: 76511-992
Supplier:  Genscript
CA125, known as cancer antigen 125, carcinoma antigen 125, and carbohydrate antigen 125, is a member of the mucin family glycoproteins. It is involved in boosting tumorigenesis and tumor proliferation and is considered as a useful biomarker for the diagnosis of ovarian cancer, Size: 10mg
Catalog #: 76511-992
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.