ContactUsLinkComponent
Pepsinogens, aspartic proteinases, are classified into two groups, pepsinogen I (PGI) and pepsinogen II (PGII). PGI and PGII are mainly secreted by gastric chief cells. They are useful in diagnosing severe atrophic gastritis and stomach cancer, Size: 100mg
Catalog #: 76511-976
Supplier:  Genscript
Pepsinogens, aspartic proteinases, are classified into two groups, pepsinogen I (PGI) and pepsinogen II (PGII). PGI and PGII are mainly secreted by gastric chief cells. They are useful in diagnosing severe atrophic gastritis and stomach cancer, Size: 100mg
Catalog #: 76511-976
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.