ContactUsLinkComponent
IgE type of immunoglobulins produced by immune system. It is composed of two heavy chains two light chains, like Ig molecules. IgE plays an essential role in various allergic reactions. Its main function is immunity to parasites such as Schistosoma mansoni, Trichinella spiralis, Fas, Size: 10mg
Catalog #: 76511-938
Supplier:  Genscript
IgE type of immunoglobulins produced by immune system. It is composed of two heavy chains two light chains, like Ig molecules. IgE plays an essential role in various allergic reactions. Its main function is immunity to parasites such as Schistosoma mansoni, Trichinella spiralis, Fas, Size: 10mg
Catalog #: 76511-938
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.