ContactUsLinkComponent
It type I transmembrane glycoprotein that belongs to B7 family. PD-L1 expressed on T cells, B cells, NK cells, dendritic cells, IFN-Y activated endothelial cells, monocytes. It plays a role in regulating T cell responses. Interaction of PD-L1 with PD-1 inhibits T cell proliferation, Size: 10mg
Catalog #: 76511-774
Supplier:  Genscript
It type I transmembrane glycoprotein that belongs to B7 family. PD-L1 expressed on T cells, B cells, NK cells, dendritic cells, IFN-Y activated endothelial cells, monocytes. It plays a role in regulating T cell responses. Interaction of PD-L1 with PD-1 inhibits T cell proliferation, Size: 10mg
Catalog #: 76511-774
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.