ContactUsLinkComponent
Programmed Cell Death Protein 1 (PD-1), is cell surface receptor expressing on T cells and pro-B cells. Binding of PD-1 to its two ligands, PD-L1 and PD-L2, could result in down-regulation of immune system by inhibiting T-cell activation process. Thus, PD-1 is an important immune checkpo, Size: 10mg
Catalog #: 76511-750
Supplier:  Genscript
Programmed Cell Death Protein 1 (PD-1), is cell surface receptor expressing on T cells and pro-B cells. Binding of PD-1 to its two ligands, PD-L1 and PD-L2, could result in down-regulation of immune system by inhibiting T-cell activation process. Thus, PD-1 is an important immune checkpo, Size: 10mg
Catalog #: 76511-750
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.