ContactUsLinkComponent
Serum Amyloid A (SAA) is an acute-phase protein. Concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs. It is considered as an early biomarker for diagnosis of many inflammatory diseases. SAA can also be used as an important indicator of infection, Size: 1mg
Catalog #: 76511-732
Supplier:  Genscript
Serum Amyloid A (SAA) is an acute-phase protein. Concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs. It is considered as an early biomarker for diagnosis of many inflammatory diseases. SAA can also be used as an important indicator of infection, Size: 1mg
Catalog #: 76511-732
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.