ContactUsLinkComponent
PCT 116 amino acid (aa) protein. It is comprised of three sections of a 57 aa N-terminal PCT, a 32 aa calcitonin a 21 aa katacalcin. Calcitonin hormone, derived from PCT cleavage. PCT good diagnosis marker for bacterial infection. Other diseases such as sepsis, inflammation, surge, Size: 1mg
Catalog #: 76511-708
Supplier:  Genscript
PCT 116 amino acid (aa) protein. It is comprised of three sections of a 57 aa N-terminal PCT, a 32 aa calcitonin a 21 aa katacalcin. Calcitonin hormone, derived from PCT cleavage. PCT good diagnosis marker for bacterial infection. Other diseases such as sepsis, inflammation, surge, Size: 1mg
Catalog #: 76511-708
Supplier:  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.